Anti-TRMT11 Rabbit Polyclonal Antibody
Catalog # AVIVARP48526_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tRNA methyltransferase 11 homolog
- Antigen symbol:TRMT11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:60487
- Antigen synonyms:TRMT11-1|C6orf75|MDS024|TRM11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TRMT11 (tRNA methyltransferase 11 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: