Anti-FNTA Rabbit Polyclonal Antibody
Catalog # AVIVARP48521_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Farnesyltransferase, CAAX box, alpha
- Antigen symbol:FNTA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:2339
- Antigen synonyms:FPTA|PGGT1A|PTAR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FNTA (farnesyltransferase, CAAX box, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: