Anti-HS3ST6 Rabbit Polyclonal Antibody
Catalog # AVIVARP48502_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:heparan sulfate (glucosamine) 3-O-sulfotransferase 6
- Antigen symbol:HS3ST6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:64711
- Antigen synonyms:h3-OST-6|3-OST-6|HS3ST5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HS3ST6 (heparan sulfate (glucosamine) 3-O-sulfotransferase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: