Anti-GSTM5 Rabbit Polyclonal Antibody
Catalog # AVIVARP48485_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:Glutathione S-transferase mu 5
- Antigen symbol:GSTM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2949
- Antigen synonyms:GSTM5-5|GTM5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: GSTM5 (glutathione S-transferase mu 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat