Anti-OXCT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48481_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:3-oxoacid CoA transferase 1
- Antigen symbol:OXCT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:5019
- Antigen synonyms:OXCT|SCOT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OXCT1 (3-oxoacid CoA transferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: