Order Entry
Sweden
ContactUsLinkComponent
Anti-OXCT1 Rabbit Polyclonal Antibody
Anti-OXCT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48480_P050
Anti-OXCT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48480_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3-oxoacid CoA transferase 1
  • Antigen symbol:
    OXCT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    5019
  • Antigen synonyms:
    OXCT|SCOT
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: OXCT1 (3-oxoacid CoA transferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: