Anti-HSPA8 Rabbit Polyclonal Antibody
Catalog # AVIVARP48446_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 8
- Antigen symbol:HSPA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3312
- Antigen synonyms:HSP73|HEL-S-72p|HSC70|LAP-1|HSC71|NIP71|HSPA10|HEL-33|HSC54|HSP71|LAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: