Anti-NUDCD1 Rabbit Polyclonal Antibody
AVIVARP48426_P050
- Antikroppstyp:Primär
- Antikroppsnamn:NudC domain containing 1
- Antigensymbol:NUDCD1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:84955
- Antigen synonymer:CML66|OVA66
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: NUDCD1 (NudC domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: