Anti-ZNF154 Rabbit Polyclonal Antibody
AVIVARP47779_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 154
- Antigen symbol:ZNF154
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Horse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7710
- Antigen synonyms:pHZ-92
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF154 (zinc finger protein 154)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Horse