Anti-SCAND3 Rabbit Polyclonal Antibody
Catalog # AVIVARP47706_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, BED-type containing 9
- Antigen symbol:ZBED9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Horse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:114821
- Antigen synonyms:SCAND3|ZNF305P2|ZFP38-L|dJ1186N24.3|Buster4|ZNF452
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SCAND3 (SCAN domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Horse