Anti-MAML3 Rabbit Polyclonal Antibody
Catalog # AVIVARP47640_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:mastermind-like transcriptional coactivator 3
- Antigen symbol:MAML3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:55534
- Antigen synonyms:MAM-2|GDN|ERDA3|MAM2|CAGH3|TNRC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: MAML3 (mastermind-like 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: