Anti-NCKAP1L Rabbit Polyclonal Antibody
AVIVARP47584_P050
- Antibody type:Primary
- Antigen name:NCK-associated protein 1-like
- Antigen symbol:NCKAP1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3071
- Antigen synonyms:HEM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NCKAP1L (NCK-associated protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: