Anti-FAR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP47392_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:Fatty Acyl CoA Reductase 2
- Antigen symbol:FAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55711
- Antigen synonyms:SDR10E2|MLSTD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: FAR2 (fatty acyl CoA reductase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: