Anti-FRRS1L Rabbit Polyclonal Antibody
AVIVARP47007_P050
- Antibody type:Primary
- Antigen name:Ferric-chelate reductase 1-like
- Antigen symbol:FRRS1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:23732
- Antigen synonyms:CG-6|C9orf4|CG6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FRRS1L (ferric-chelate reductase 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: