Anti-SWAP70 Rabbit Polyclonal Antibody
Catalog # AVIVARP46214_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SWAP switching B-cell complex 70kDa subunit
- Antigen symbol:SWAP70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:23075
- Antigen synonyms:HSPC321|SWAP-70
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SWAP70 (SWAP switching B-cell complex 70kDa subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: