Anti-HSPA4 Rabbit Polyclonal Antibody
AVIVARP46073_P050
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 4
- Antigen symbol:HSPA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3308
- Antigen synonyms:HS24/P52|HSPH2|APG-2|HEL-S-5a|hsp70|hsp70RY|RY
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HSPA4 (heat shock 70kDa protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: