Anti-KLRC3 Rabbit Polyclonal Antibody
Catalog # AVIVARP45260_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:killer cell lectin-like receptor subfamily C, member 3
- Antigen symbol:KLRC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3823
- Antigen synonyms:NKG2-E|NKG2E
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: KLRC3 (killer cell lectin-like receptor subfamily C, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human