Anti-MEGF6 Rabbit Polyclonal Antibody
Catalog # AVIVARP45090_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:multiple EGF-like-domains 6
- Antigen symbol:MEGF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:65049
- Antigen synonyms:Egfl3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MEGF6 (multiple EGF-like-domains 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Rat