Anti-DSCAM Rabbit Polyclonal Antibody
AVIVARP45086_P050
- Antibody type:Primary
- Antigen name:Down syndrome cell adhesion molecule
- Antigen symbol:DSCAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1826
- Antigen synonyms:CHD2-42|CHD2|CHD2-52
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DSCAM (Down syndrome cell adhesion molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: