Anti-AGPAT2 Rabbit Polyclonal Antibody
Catalog # AVIVARP44636_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:1-Acylglycerol-3-phosphate O-acyltransferase 2
- Antigen symbol:AGPAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10555
- Antigen synonyms:LPAAB|BSCL|1-AGPAT2|LPAAT-beta|BSCL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AGPAT2 (1-acylglycerol-3-phosphate O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: