Anti-B4GALT2 Rabbit Polyclonal Antibody
Catalog # AVIVARP44507_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2
- Antigen symbol:B4GALT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:8704
- Antigen synonyms:beta4Gal-T2|B4Gal-T2|B4Gal-T3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: B4GALT2 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: