Anti-LRRC52 Rabbit Polyclonal Antibody
Katalog # AVIVARP44494_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:leucine rich repeat containing 52
- Antigen symbol:LRRC52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:440699
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: LRRC52 (leucine rich repeat containing 52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: