Anti-SLC5A4 Rabbit Polyclonal Antibody
AVIVARP43941_P050
- Antibody type:Primary
- Antigen name:solute carrier family 5 (glucose activated ion channel), member 4
- Antigen symbol:Solute carrier family 5, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6527
- Antigen synonyms:SAAT1|SGLT3|DJ90G24.4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC5A4 (solute carrier family 5 (glucose activated ion channel), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: