Anti-MARCH1 Rabbit Polyclonal Antibody
Catalog # AVIVARP43239_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Membrane associated ring-CH-type finger 1
- Antigen symbol:MARCH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55016
- Antigen synonyms:MARCH-I|RNF171
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MARCH1 (membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat