Anti-MKRN1 Rabbit Polyclonal Antibody
AVIVARP43142_T100
- Antibody type:Primary
- Antigen name:makorin ring finger protein 1
- Antigen symbol:MKRN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23608
- Antigen synonyms:RNF61
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: MKRN1 (makorin ring finger protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: