Anti-WWP2 Rabbit Polyclonal Antibody
Katalog # AVIVARP43088_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:WW domain containing E3 ubiquitin protein ligase 2
- Antigen symbol:WWP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:11060
- Antigen synonyms:WWp2-like|AIP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: WWP2 (WW domain containing E3 ubiquitin protein ligase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: