Anti-FTCD Rabbit Polyclonal Antibody
Catalog # AVIVARP41576_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Formimidoyltransferase Cyclodeaminase
- Antigen symbol:FTCD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10841
- Antigen synonyms:LCHC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: