Anti-PRPF6 Rabbit Polyclonal Antibody
Catalog # AVIVARP40735_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:pre-mRNA processing factor 6
- Antigen symbol:PRPF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:24148
- Antigen synonyms:SNRNP102|hPrp6|U5-102K|RP60|Prp6|C20orf14|ANT1|TOM|ANT-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PRPF6 (pre-mRNA processing factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: