Anti-NONO Rabbit Polyclonal Antibody
Catalog # AVIVARP40716_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Non-POU domain containing, octamer-binding
- Antigen symbol:NONO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4841
- Antigen synonyms:P54|NRB54|P54NRB|NMT55|PPP1R114
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NONO (non-POU domain containing, octamer-binding)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: