Anti-TADA3 Rabbit Polyclonal Antibody
Catalog # AVIVARP38786_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transcriptional adaptor 3
- Antigen symbol:TADA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10474
- Antigen synonyms:ADA3|TADA3L|hADA3|STAF54|NGG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TADA3 (transcriptional adaptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Zebrafish