Order Entry
Sweden
ContactUsLinkComponent
Anti-ALX3 Rabbit Polyclonal Antibody
Anti-ALX3 Rabbit Polyclonal Antibody
  AVIVARP37852_T100
undefined
Anti-ALX3 Rabbit Polyclonal Antibody
  AVIVARP37852_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    Aristaless-like homeobox 3
  • Antigen symbol:
    ALX3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    11694
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: ALX3 (ALX homeobox 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat