Anti-NKX1-1 Rabbit Polyclonal Antibody
AVIVARP37638_P050
- Antibody type:Primary
- Antigen name:NK1 homeobox 1
- Antigen symbol:NKX1-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:20232
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NKX1-1 (NK1 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse