Anti-FOXQ1 Rabbit Polyclonal Antibody
AVIVARP36873_P050
- Antikroppstyp:Primär
- Antikroppsnamn:Forkhead Box Q1
- Antigensymbol:FOXQ1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Rat,Mouse
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:15220
- Antigen synonymer:Hfh1l|HFH-1|Hfh1|sa
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: FOXQ1 (forkhead box Q1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat