Order Entry
Sweden
ContactUsLinkComponent
Anti-ZNF471 Rabbit Polyclonal Antibody
Anti-ZNF471 Rabbit Polyclonal Antibody
  AVIVARP35981_P050
Anti-ZNF471 Rabbit Polyclonal Antibody
  AVIVARP35981_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger protein 471
  • Antigen symbol:
    ZNF471
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    57573
  • Antigen synonyms:
    ERP1|Z1971
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: ZNF471 (zinc finger protein 471)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Pig