Anti-MESP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP35960_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Mesoderm posterior 1 homolog
- Antigen symbol:MESP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55897
- Antigen synonyms:bHLHc5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MESP1 (mesoderm posterior 1 homolog (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human