Anti-ASIC2 Rabbit Polyclonal Antibody
Catalog # AVIVARP35537_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Acid sensing (proton-gated) ion channel 2
- Antigen symbol:ASIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:40
- Antigen synonyms:MDEG|BNaC1|ASIC2a|hBNaC1|ACCN|ACCN1|BNC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ASIC2 (acid-sensing (proton-gated) ion channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: