Anti-MCOLN3 Rabbit Polyclonal Antibody
Catalog # AVIVARP35278_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Mucolipin 3
- Antigensymbol:MCOLN3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:55283
- Antigen synonymer:TRPML3|TRP-ML3
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: MCOLN3 (mucolipin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: