Anti-KCNN2 Rabbit Polyclonal Antibody
Catalog # AVIVARP35094_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 2
- Antigen symbol:KCNN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3781
- Antigen synonyms:hSK2|SKCA2|SK2|SKCa 2|KCa2.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KCNN2 (potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: