Order Entry
Sweden
ContactUsLinkComponent
Anti-SCN5A Rabbit Polyclonal Antibody
Anti-SCN5A Rabbit Polyclonal Antibody
  AVIVARP34934_P050
Anti-SCN5A Rabbit Polyclonal Antibody
  AVIVARP34934_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, voltage gated, type V alpha subunit
  • Antigen symbol:
    SCN5A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    6331
  • Antigen synonyms:
    Nav1.5|ICCD|CMPD2|HH1|VF1|CMD1E|SSS1|IVF|CDCD2|HB2|HB1|PFHB1|HBBD|LQT3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: SCN5A (sodium channel, voltage-gated, type V, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: