Order Entry
Sweden
ContactUsLinkComponent
Anti-PHF11 Rabbit Polyclonal Antibody
Anti-PHF11 Rabbit Polyclonal Antibody
Catalog # AVIVARP34848_P050
Anti-PHF11 Rabbit Polyclonal Antibody
Catalog # AVIVARP34848_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PHD finger protein 11
  • Antigen symbol:
    PHF11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    51131
  • Antigen synonyms:
    APY|IGEL|NY-REN-34|NYREN34|IGHER|IGER|BCAP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: PHF11 (PHD finger protein 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: