Anti-WBP5 Rabbit Polyclonal Antibody
AVIVARP34454_P050
- Antibody type:Primary
- Antigen name:WW domain binding protein 5
- Antigen symbol:WBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:51186
- Antigen synonyms:TCEAL9|WEX6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: WBP5 (WW domain binding protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: