Anti-ZFP36 Rabbit Polyclonal Antibody
Catalog # AVIVARP34385_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:ZFP36 ring finger protein
- Antigen symbol:ZFP36
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7538
- Antigen synonyms:RNF162A|NUP475|zfp-36|TTP|G0S24|GOS24|TIS11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: ZFP36 (ZFP36 ring finger protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: