Order Entry
Sweden
ContactUsLinkComponent
Anti-CTRC Rabbit Polyclonal Antibody
Anti-CTRC Rabbit Polyclonal Antibody
  AVIVARP33873_P050
undefined
Anti-CTRC Rabbit Polyclonal Antibody
  AVIVARP33873_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    chymotrypsin C (caldecrin)
  • Antigen symbol:
    CTRC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    11330
  • Antigen synonyms:
    ELA4|CLCR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: CTRC (chymotrypsin C (caldecrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat