Anti-GZF1 Rabbit Polyclonal Antibody
AVIVARP33281_P050
- Antikroppstyp:Primär
- Antikroppsnamn:GDNF-inducible zinc finger protein 1
- Antigensymbol:GZF1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:64412
- Antigen synonymer:ZNF336|ZBTB23
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: GZF1 (GDNF-inducible zinc finger protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat