Anti-PIAS3 Rabbit Polyclonal Antibody
Catalog # AVIVARP32762_T100
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Protein inhibitor of activated STAT 3
- Antigensymbol:PIAS3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:10401
- Antigen synonymer:ZMIZ5
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:100 µG
Specifications
About this item
Type: Primary
Antigen: PIAS3 (protein inhibitor of activated STAT, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: