Anti-PSMD14 Rabbit Polyclonal Antibody
AVIVARP32450_T100
- Antibody type:Primary
- Antigen name:proteasome 26S subunit, non-ATPase 14
- Antigen symbol:PSMD14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10213
- Antigen synonyms:PAD1|RPN11|POH1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: PSMD14 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: