Anti-ELF4 Rabbit Polyclonal Antibody
AVIVARP31640_P050
- Antibody type:Primary
- Antigen name:E74-like factor 4 (ets domain transcription factor)
- Antigen symbol:ELF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2000
- Antigen synonyms:ELFR|MEF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ELF4 (E74-like factor 4 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: