Anti-ATRAID Rabbit Polyclonal Antibody
AVIVARP30550_P050
- Antibody type:Primary
- Antigen name:all-trans retinoic acid-induced differentiation factor
- Antigen symbol:ATRAID
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51374
- Antigen synonyms:APR--3|HSPC013|PRO240|APR-3|C2orf28|p18|APR3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ATRAID (all-trans retinoic acid-induced differentiation factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: