Anti-AMH Rabbit Polyclonal Antibody
Catalog # BOSSBS-4687R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Anti-Mullerian Hormone
- Antigen symbol:AMH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:anti-Mullerian hormone|MIF|MIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: AMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat