Anti-ADAM9 Rabbit Polyclonal Antibody
Catalog # BOSSBS-4204R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 9
- Antigen symbol:ADAM9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Antigen synonyms:ADAM9|KIAA0021|Metalloprotease/disintegrin/cysteine-rich protein 9|Meltrin-gamma|MLTNG|Cellular disintegrin-related protein|A disintegrin and metalloproteinase domain 9|ADAM 9|Myeloma cell metalloproteinase|MCMP|MDC9
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ADAM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat